missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyaniline, 98%

Product Code. 11467367
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467367 50 g 50g
11457367 10 g 10g
11369519 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467367 Supplier Thermo Scientific Alfa Aesar Supplier No. A15073.18
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10272-07-8 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10272-07-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008392
InChI Key WNRGWPVJGDABME-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423
PubChem CID 66301
IUPAC Name 3,5-dimethoxyaniline
SMILES COC1=CC(OC)=CC(N)=C1

Specifications

Melting Point 53°C to 57°C
Boiling Point 178°C (20 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 50 g
UN Number UN2811
Beilstein 638013
Sensitivity Light sensitive
Formula Weight 153.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.