missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyaniline, 98%

Product Code. 10160452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160452 5 g Glass bottle 5g
10570731 25 g Glass bottle 25g
10580731 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10160452 Supplier Thermo Scientific Acros Supplier No. 115360050
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10272-07-8 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10272-07-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008392
InChI Key WNRGWPVJGDABME-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423
PubChem CID 66301
IUPAC Name 3,5-dimethoxyaniline
SMILES COC1=CC(OC)=CC(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 53.0°C to 55.0°C
Color Brown to Colorless
Boiling Point 178.0°C (20.0 mmHg)
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3NH2
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 153.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 5-Dimethoxyaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.