missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl bromide, 95%

Product Code. 10694545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694545 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10694545 Supplier Thermo Scientific Acros Supplier No. 427340050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 877-88-3 | C9H11BrO2 | 231.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 877-88-3
CAS Min % 94.0
CAS Max % 100.0
Melting Point 69°C to 70°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H11BrO2
Linear Formula (CH30)2C6H3CH2Br
MDL Number MFCD01321368
Quantity 5 g
Synonym 3,5-dimethoxybenzyl bromide, 1-bromomethyl-3,5-dimethoxybenzene, 3,5-dimethoxybenzylbromide, benzene, 1-bromomethyl-3,5-dimethoxy, 3,5-dimethoxy benzyl bromide, alpha-bromo-3,5-dimethoxytoluene, 5-bromomethyl-1,3-dimethoxybenzene, 5-bromomethyl resorcinol dimethyl ether, pubchem13588, acmc-209qq1
InChI Key BTHIGJGJAPYFSJ-UHFFFAOYSA-N
SMILES COC1=CC(=CC(=C1)CBr)OC
IUPAC Name 1-(bromomethyl)-3,5-dimethoxybenzene
Molecular Weight (g/mol) 231.08
PubChem CID 1274490
Formula Weight 231.08
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dimethoxybenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethoxybenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.