missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenol, 97%

Product Code. 10341474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10341474 50 g Glass bottle 50g
10235680 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10341474 Fornitore Thermo Scientific Acros N. del fornitore 158120500
 Restricted Item
Estimated Shipment: 07-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 500-99-2 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 500-99-2
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008388
InChI Key XQDNFAMOIPNVES-UHFFFAOYSA-N
Synonym taxicatigenin, phenol, 3,5-dimethoxy, phloroglucinol dimethyl ether, 1-hydroxy-3,5-dimethoxybenzene, 3,5-dimethoxy phenol, unii-23uxw8136a, phenol,5-dimethoxy, 3,5-dimethoxy-phenol, pubchem16421, acmc-209kk2
PubChem CID 10383
IUPAC Name 3,5-dimethoxyphenol
SMILES COC1=CC(OC)=CC(O)=C1

Specifica

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color Brown-Pink to Colorless
Boiling Point 172°C to 175°C (17.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3OH
Quantity 50 g
Beilstein 06, 1101
Formula Weight 154.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Dimethoxyphenol
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 3,5-Dimethoxyphenol
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.