missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxytoluene, 99%, Thermo Scientific™

Product Code. 10504334 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
1g; Glass bottle
This item is not returnable. View return policy

Product Code. 10504334

Brand: Thermo Scientific Acros 391190010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4179-19-5
CAS Min % 98.5
CAS Max % 100.0
Density 1.0390g/mL
Boiling Point 244.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O2
Linear Formula CH3C6H3(OCH3)2
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
MDL Number MFCD00015435
Quantity 1g
Specific Gravity 1.039
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5i
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
SMILES COC1=CC(OC)=CC(C)=C1
IUPAC Name 1,3-dimethoxy-5-methylbenzene
Molecular Weight (g/mol) 152.19
PubChem CID 77844
Formula Weight 152.19
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethoxytoluene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.