missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-hydroxybenzonitrile, 97%, Thermo Scientific™

Produktkod. 10081851 Handla allt Thermo Scientific Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod.
10081851 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10081851 Leverantör Thermo Scientific Acros Leverantörsnummer 353660050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Specifikationer

CAS 4198-90-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.0°C to 126.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NO
MDL Number MFCD00221716
Quantity 5 g
Beilstein 10, IV, 681
Synonym 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
InChI Key WFYGXOWFEIOHCZ-UHFFFAOYSA-N
SMILES CC1=CC(=CC(=C1O)C)C#N
IUPAC Name 4-hydroxy-3,5-dimethylbenzonitrile
Molecular Weight (g/mol) 147.18
PubChem CID 20176
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dimethyl-4-hydroxybenzonitrile
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 3,5-Dimethyl-4-hydroxybenzonitrile
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.