missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-nitrobenzoic acid, 97%

Product Code. 11387795
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387795 1 g 1g
11397795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387795 Supplier Thermo Scientific Alfa Aesar Supplier No. L02704.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3095-38-3 | C9H9NO4 | 195.17 g/mol

3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3095-38-3
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00014699
InChI Key RBAVFNOGEPCOQI-UHFFFAOYSA-N
PubChem CID 249413
IUPAC Name 3,5-dimethyl-4-nitrobenzoic acid
SMILES CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O

Specifications

Melting Point 222°C to 226°C
Odor Odorless
Quantity 1 g
Beilstein 1965772
Synonym 3,5-dimethyl-4-nitrobenzoic acid, benzoic acid, 3,5-dimethyl-4-nitro, 4-nitro-3,5-dimethylbenzoic acid, 3,5-dimethyl-4-nitro-benzoic acid, 4-nitromesitylenic acid, pubchem19635, acmc-1agmi, benzoic acid,5-dimethyl-4-nitro, benzoic acid,3,5-dimethyl-4-nitro, 3,5-dimethyl-4-nitro benzoic acid
Solubility Information Insoluble in water.
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-4-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.