missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4H-1,2,4-triazol-4-amine, 97%, Thermo Scientific™

Product Code. 10477042
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477042 1 g Amber Glass Bottle 1g
10792673 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10477042 Supplier Thermo Scientific Maybridge Supplier No. CD08995DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3530-15-2
Molecular Formula C4H8N4
Molecular Weight (g/mol) 112.136
MDL Number MFCD00051647
InChI Key MIIKMZAVLKMOFM-UHFFFAOYSA-N
PubChem CID 229800
IUPAC Name 3,5-dimethyl-1,2,4-triazol-4-amine
SMILES CC1=NN=C(N1N)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3,5-dimethyl-4h-1,2,4-triazol-4-amine, 4-amino-3,5-dimethyl-1,2,4-triazole, dimethyl-1,2,4-triazol-4-amine, dimethyl-4h-1,2,4-triazol-4-amine, 4h-1,2,4-triazol-4-amine, 3,5-dimethyl, 3,5-dimethyl-1,2,4-triazole-4-ylamine, acmc-1cpm8, 3,5-dimethyl-4-amino-1,2,4-triazole, 4-amino-3,5-dimethyl-1,2,4 triazole, 4h-1,2,4-triazol-4-amine,3,5-dimethyl
Formula Weight 112.14
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-4H-1,2,4-triazol-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.