missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol, 98+%

Product Code. p-7082008
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11474923

Brand: Thermo Scientific Alfa Aesar A12236.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-68-9 | C8H10O | 122.17 g/mol

3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).

Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.

Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

Melting Point 61°C to 65°C
Density 1.12
Boiling Point 222°C
Flash Point 109°C (228°F)
Quantity 100 g
UN Number UN2261
Beilstein 774117
Merck Index 14,10082
Solubility Information Soluble in ethyl alcohol,carbon tetrachloride,benzene,chloroform,water and ether.
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylphenol
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.