missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenol, 99+%

Product Code. 10194460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194460 5 g Glass bottle 5g
10225880 100 g Glass bottle 100g
10696942 500 g Glass bottle 500g
10524524 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10194460 Supplier Thermo Scientific Acros Supplier No. 162790050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-68-9 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-68-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002307
InChI Key TUAMRELNJMMDMT-UHFFFAOYSA-N
Synonym 3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa
PubChem CID 7948
ChEBI CHEBI:38572
IUPAC Name 3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63°C to 66°C
Color Orange to White
Density 1.1150g/mL
Boiling Point 222°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3OH
Quantity 5 g
Beilstein 06, 492
Merck Index 15, 10279
Specific Gravity 1.115
Solubility Information Solubility in water: 5.3g/L (25°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Percent Purity 99+%
Physical Form Crystalline Solid
Chemical Name or Material 3, 5-Dimethylphenol, 99+%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.