missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoic acid, 98+%

Product Code. 13448378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13448378 1 kg Plastic bottle 1kg
10387370 100 g Plastic bottle 100g
10153010 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13448378 Supplier Thermo Scientific Acros Supplier No. 153190010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-34-3
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007253
InChI Key VYWYYJYRVSBHJQ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
PubChem CID 7433
ChEBI CHEBI:73914
IUPAC Name 3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Density 1.6830g/mL
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Linear Formula (O2N)2C6H3CO2H
Quantity 1 kg
Beilstein 09,413
Merck Index 15,3302
Specific Gravity 1.683
Solubility Information Solubility in water: 1350mg/l (25°C). Other solubilities: very soluble in alcohol and glacial acetic acid,sparingly soluble in ether,benzene and,carbon disulfide
Formula Weight 212.12
Percent Purity 98+%
Chemical Name or Material 3, 5-Dinitrobenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dinitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.