missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylic acid, 97+%

Product Code. 11488826
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488826 50 g 50g
11498826 250 g 250g
11478826 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488826 Supplier Thermo Scientific Alfa Aesar Supplier No. A16988.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 609-99-4 | C7H4N2O7 | 228.116 g/mol

3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.

Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-99-4
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.116
MDL Number MFCD00007104
InChI Key LWFUFLREGJMOIZ-UHFFFAOYSA-N
Synonym 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid
PubChem CID 11873
ChEBI CHEBI:53648
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 169°C to 174°C
Quantity 50 g
Beilstein 2220661
Solubility Information Soluble in water,dimethyl sulfoxide,methanol,ethanol,diethyl ether and benzene.
Formula Weight 228.12
Percent Purity ≥97%
Chemical Name or Material 3,5-Dinitrosalicylic acid
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dinitrosalicylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Dinitrosalicylicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.