missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylic acid, 98%

Product Code. 10664632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10664632 500 g Glass bottle 500g
10654632 100 g Glass bottle 100g
15139203 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10664632 Supplier Thermo Scientific Acros Supplier No. 156445000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3,5-dinitrosalicylic acid, CAS # 609-99-4, also known as DNS or DNSA, is used for quantifying carbohydrate levels in blood by calorimetry. | CAS: 609-99-4 | C7H4N2O7 | 228.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

3,5-Dinitrosalicylic acid also known as DNA or DNSA reacts with reducing sugars forming 3-amino-5-nitrosalicylic acid, which is an aromatic compound with maximum absorption at 540 nm allowing spectrophotometer measurement of the amount of reducing molecules

Applications

  • It is used in the quantification of reducing sugars by colorimetry in the blood. It is also used to analyze glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar
  • 3,5-Dinitrosalicylic acid can detect reducing substances in the urine
  • This compound is used to detect the presence of free carbonyl groups (C=O)
  • 3,5-Dinitrosalicylic acid can be used to determine the effects of silver nanoparticles (AgNPs) on membrane leakage
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-99-4
Molecular Formula C7H4N2O7
Molecular Weight (g/mol) 228.12
MDL Number MFCD00007104
InChI Key LWFUFLREGJMOIZ-UHFFFAOYSA-N
Synonym 3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid
PubChem CID 11873
ChEBI CHEBI:53648
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 170.0°C to 174.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (O2N)2C6H2(OH)CO2H
Quantity 500 g
Beilstein 10,122
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and benzene
Formula Weight 228.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3,5-Dinitrosalicylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dinitrosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.