missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diacetoxy Fluorescein Bis(methylene)bis(azanetriyl)tetraacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3,6-Diacetoxy Fluorescein Bis(methylene)bis(azanetriyl)tetraacetic Acid |
| Analyte or Component Names | 3,6-Diacetoxy Fluorescein Bis(methylene)bis(azanetriyl)tetraacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H30 N2 O15 |
| InChI Formula | InChI=1S/C34H30N2O15/c1-17(37)48-25-9-7-23-31(20(25)11-35(13-27(39)40)14-28(41)42)50-32-21(12-36(15-29(43)44)16-30(45)46)26(49-18(2)38)10-8-24(32)34(23)22-6-4-3-5-19(22)33(47)51-34/h3-10H,11-16H2,1-2H3,(H,39,40)(H,41,42)(H,43,44)(H,45,46) |
| Solution Type | Neat |
| SMILES | CC(=O)Oc1ccc2c(Oc3c(CN(CC(=O)O)CC(=O)O)c(OC(=O)C)ccc3C24OC(=O)c5ccccc45)c1CN(CC(=O)O)CC(=O)O |
| IUPAC Name | 2-[carboxymethyl-[[3',6'-diacetyloxy-5'-[[bis(carboxymethyl)amino]methyl]-3-oxospiro[2-benzofuran-1,9'-xanthene]-4'-yl]methyl]amino]acetic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 706.606 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?