missing translation for 'onlineSavingsMsg'
Learn More

3-[(6-Methylpyrazin-2-yl)oxy]benzylamine, 97%, Thermo Scientific™

Product Code. 10326764
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10326764 250 mg Amber Glass Bottle 250mg
10505754 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326764 Supplier Thermo Scientific Maybridge Supplier No. CC62613CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 941716-89-8
Molecular Formula C12H13N3O
Molecular Weight (g/mol) 215.256
MDL Number MFCD09817526
InChI Key PJVRJIDLKXDROC-UHFFFAOYSA-N
Synonym 3-6-methylpyrazin-2-yl oxy benzylamine, 3-6-methylpyrazin-2-yl oxy phenyl methanamine, 1-3-6-methylpyrazin-2-yl oxy phenyl methanamine, 3-6-methylpyrazin-2-yloxy phenyl methylamine, 3-6-methylpyrazin-2-yl oxy phenyl methylamine
PubChem CID 24229681
IUPAC Name [3-(6-methylpyrazin-2-yl)oxyphenyl]methanamine
SMILES CC1=CN=CC(=N1)OC2=CC=CC(=C2)CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.25
Percent Purity 90%
Chemical Name or Material 3-[(6-Methylpyrazin-2-yl)oxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.