missing translation for 'onlineSavingsMsg'
Learn More

3-(9-Anthryl)propionic acid, 96%

Product Code. 15417117
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417117 5 g 5g
15427117 250 mg 250mg
15407117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15417117 Supplier Thermo Scientific Alfa Aesar Supplier No. H34184.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 41034-83-7 | C17H14O2 | 250.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41034-83-7
Molecular Formula C17H14O2
Molecular Weight (g/mol) 250.30
MDL Number MFCD00068641
InChI Key BDGMYCZUEIGHJH-UHFFFAOYSA-N
Synonym 9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid
PubChem CID 170457
IUPAC Name 3-anthracen-9-ylpropanoic acid
SMILES OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 193°C to 197°C
Quantity 5 g
Formula Weight 250.3
Percent Purity 96%
Chemical Name or Material 3-(9-Anthryl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.