missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidobenzeneboronic acid, 98%

Product Code. 11430308
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430308 25 g 25g
11410308 1 g 1g
11420308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430308 Supplier Thermo Scientific Alfa Aesar Supplier No. B23833.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78887-39-5 | C8H10BNO3 | 178.982 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78887-39-5
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.982
MDL Number MFCD00236013
InChI Key IBTSWKLSEOGJGJ-UHFFFAOYSA-N
Synonym 3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid
PubChem CID 157274
IUPAC Name (3-acetamidophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)NC(=O)C)(O)O

Specifications

Melting Point ∼263°C (decomposition)
Quantity 25 g
Beilstein 3278316
Formula Weight 178.98
Percent Purity 98%
Chemical Name or Material 3-Acetamidobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.