missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenol, 98%

Product Code. p-7020715
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11349519

Brand: Thermo Scientific Alfa Aesar A12764.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-42-1 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-42-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002263
InChI Key QLNWXBAGRTUKKI-UHFFFAOYSA-N
Synonym 3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
PubChem CID 12124
ChEBI CHEBI:76987
IUPAC Name N-(3-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)O

Specifications

Melting Point 146°C to 149°C
Quantity 25 g
Beilstein 907998
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Acetamidophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.