missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-2-methylbenzoic acid, 98%, Thermo Scientific™

Product Code. 11945101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11945101 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945101 Supplier Thermo Scientific Acros Supplier No. 443860050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 168899-58-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 148°C to 151°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C10H10O4
Quantity 5 g
Synonym 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid
InChI Key LHVJUPHCLWIPLZ-UHFFFAOYSA-N
SMILES CC1=C(C=CC=C1OC(=O)C)C(=O)O
IUPAC Name 3-acetyloxy-2-methylbenzoic acid
Molecular Weight (g/mol) 194.19
PubChem CID 3798557
Formula Weight 194.19
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3-Acetoxy-2-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetoxy-2-methyl-benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.