missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-2-methylbenzoic acid, 99%, Thermo Scientific™

Product Code. 11349495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349495 25 g 25g
11339495 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349495 Supplier Thermo Scientific Alfa Aesar Supplier No. L18719.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 168899-58-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00957176
InChI Key LHVJUPHCLWIPLZ-UHFFFAOYSA-N
Synonym 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid
PubChem CID 3798557
IUPAC Name 3-acetyloxy-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1OC(=O)C)C(=O)O

Specifikationer

Melting Point 149°C to 150°C
Quantity 25 g
Beilstein 3271605
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material 3-Acetoxy-2-methylbenzoic acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.