missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-deoxynivalenol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30310942
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310942 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310942 Supplier Dr. Ehrenstorfer Supplier No. DREV10233000AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 3-Acetyl-deoxynivalenol
CAS 50722-38-8
Analyte or Component Names 3-Acetyl-deoxynivalenol
Shipping Condition Room Temperature
Molecular Formula C17 H22 O7
Concentration 100 μg/mL
Certifications/Compliance ISO 9001
Synonym (3α,7α)-3-(Acetyloxy)-12,13-epoxy-7,15-dihydroxytrichothec-9-en-8-one, Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane], trichothec-9-en-8-one deriv., 3-Acetyldeoxynivalenol, 3α-Acetoxy-12,13-epoxytrichothec-9-en-8-one, DON 3-acetate, Dehydronivalenol monoacetate, Deoxynivalenol 3-acetate, Deoxynivalenol monoacetate, Monoacetyldeoxynivalenol, NSC 267036
InChI Formula InChI=1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1
Solution Type Single Solution
SMILES CC(=O)O[C@@H]1C[C@@]2(C)[C@]3(CO3)[C@@H]1O[C@@H]4C=C(C)C(=O)[C@@H](O)[C@]24CO
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 338.35
Quantity 5 mL
Formula Weight 338.1366 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.