missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylcoumarin, 98+%

Product Code. 11424204
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424204 25 g 25g
11414204 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424204 Supplier Thermo Scientific Alfa Aesar Supplier No. A12532.14
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3949-36-8
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00006853
InChI Key CSPIFKKOBWYOEX-UHFFFAOYSA-N
Synonym 3-acetylcoumarin, 3-acetyl-2h-chromen-2-one, coumarin, 3-acetyl, 2h-1-benzopyran-2-one, 3-acetyl, 3-acetyl-2-oxochromene, 3-acetylcoumarine, pubchem8664, maybridge1_000885, prestwick0_000891, prestwick1_000891
PubChem CID 77553
IUPAC Name 3-acetylchromen-2-one
SMILES CC(=O)C1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 119°C to 124°C
Quantity 25 g
Beilstein 143498
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material 3-Acetylcoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.