missing translation for 'onlineSavingsMsg'
Learn More

3-{[(Allyloxy)carbonyl]amino}-5-hydroxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11386895
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11386895 5 g Amber Glass Bottle 5g
10592594 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386895 Supplier Thermo Scientific Maybridge Supplier No. MO08020DE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 916766-99-9
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.21
MDL Number MFCD09702421
InChI Key MYOCYAGTJQROGX-UHFFFAOYSA-N
Synonym 3-allyloxy carbonyl amino-5-hydroxybenzoic acid, 3-hydroxy-5-prop-2-en-1-yloxy carbonyl amino benzoic acid, allyl 3-carboxy-5-hydroxyphenyl carbamate, 5-hydroxy-3-prop-2-enyloxycarbonylamino benzoic acid, benzoicacid,3-hydroxy-5-2-propen-1-yloxy carbonyl amino, 3-hydroxy-5-prop-2-en-1-yl oxy carbonyl amino benzoic acid, benzoicacid, 3-hydroxy-5-2-propen-1-yloxy carbonyl amino
PubChem CID 24229781
IUPAC Name 3-hydroxy-5-(prop-2-enoxycarbonylamino)benzoic acid
SMILES OC(=O)C1=CC(NC(=O)OCC=C)=CC(O)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 237.21
Percent Purity 97%
Chemical Name or Material 3-{[(allyloxy)carbonyl]amino}-5-hydroxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.