missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,4,6-triiodobenzoic acid, 99% (dry wt.), water <4%

Product Code. 11441017
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441017 25 g 25g
11431017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441017 Supplier Thermo Scientific Alfa Aesar Supplier No. B23405.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3119-15-1 | C7H3I3NNaO2 | 536.81 g/mol

3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Protect from exposure to light. Store in the dark. Incompatible with light and oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3119-15-1
Molecular Formula C7H3I3NNaO2
Molecular Weight (g/mol) 536.81
MDL Number MFCD00007681
InChI Key CTXRMWPLTDAHOR-UHFFFAOYSA-M
Synonym unii-99h77d8mtd, 3-amino-2,4,6-triiodo-benzoic acid, acido 3-amino-2,4,6-triiodobenzoico italian, benzoic acid, 3-amino-2,4,6-triiodo, 3-amino-2,4,6-triiodo-benzoicacid, acido 3-amino-2,4,6-triiodobenzoico, pubchem4987, acmc-1co9w, 4-14-00-01118 beilstein handbook reference, 2,4,6-triiodo-3-aminobenzoic acid
PubChem CID 18387
IUPAC Name 3-amino-2,4,6-triiodobenzoic acid
SMILES [Na+].NC1=C(I)C=C(I)C(C([O-])=O)=C1I

Specifications

Melting Point ∼196°C (decomposition)
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 514.83
Percent Purity 99%
Assay (dry wt.), may cont. up to 1 mole of water
Chemical Name or Material 3-Amino-2,4,6-triiodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.