missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthoic acid, 97%

Product Code. 11364226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364226 5 g 5g
11354226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364226 Supplier Thermo Scientific Alfa Aesar Supplier No. L19312.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5959-52-4 | C11H9NO2 | 187.198 g/mol

3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-52-4
Molecular Formula C11H9NO2
MDL Number MFCD00004115
InChI Key XFXOLBNQYFRSLQ-UHFFFAOYSA-N
Synonym 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
PubChem CID 22244
IUPAC Name 3-aminonaphthalene-2-carboxylic acid

Specifications

Melting Point ∽210°C (decomposition)
Quantity 5 g
Beilstein 744099
Merck Index 14,452
Solubility Information Soluble in alcohol and ether. Insoluble in water.
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N
Molecular Weight (g/mol) 187.198
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-Amino-2-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.