missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzoic acid, 98%

Product Code. 10535681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10535681 5 g 5g
10183101 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535681 Supplier Thermo Scientific Acros Supplier No. 271490050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1571-72-8 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-72-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007697
InChI Key MRBKRZAPGUCWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
PubChem CID 65083
ChEBI CHEBI:29476
IUPAC Name 3-amino-4-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 198.0°C to 205.0°C
Color Beige-Gray to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(OH)COOH
Quantity 5 g
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-hydroxybenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.