missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzanilide, 98%, Thermo Scientific™

Product Code. 11429456
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429456 50 g 50g
11419456 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429456 Supplier Thermo Scientific Alfa Aesar Supplier No. A19168.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-35-4
Molecular Formula C14H14N2O2
Molecular Weight (g/mol) 242.278
MDL Number MFCD00017166
InChI Key LHMQDVIHBXWNII-UHFFFAOYSA-N
Synonym 3-amino-4-methoxybenzanilide, 3-amino-p-anisanilide, benzamide, 3-amino-4-methoxy-n-phenyl, 3-amino-4-methoxy benzanilide, benzanilide, 3-amino-4-methoxy, p-anisanilide, 3-amino, unii-v5y646aw8s, amba, 3-amino-4-methoxy-n-phenyl-benzamide, 3-amino-4-methoxyphenyl-n-benzamide
PubChem CID 8426
IUPAC Name 3-amino-4-methoxy-N-phenylbenzamide
SMILES COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N

Specifications

Melting Point 154°C to 156°C
Quantity 50 g
Formula Weight 242.28
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methoxybenzanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.