missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzoic acid, 98+%

Product Code. p-7025274
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11449127

Brand: Thermo Scientific Alfa Aesar B20669.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2840-26-8 | C8H9NO3 | 167.164 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2840-26-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00002521
InChI Key FDGAEAYZQQCBRN-UHFFFAOYSA-N
Synonym 3-amino-p-anisic acid, p-anisic acid, 3-amino, 3-amino-4-methoxy-benzoic acid, 3-amino-4-anisic acid, 3-amino-4-methoxybenzoicacid, benzoic acid, 3-amino-4-methoxy, kyselina 3-amino-4-methoxybenzoova, 3-amino-4-methoxybenzoic, 5-carboxy-2-methoxyaniline, kyselina 3-amino-4-methoxybenzoova czech
PubChem CID 17823
IUPAC Name 3-amino-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 208°C to 210°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 167.16
Percent Purity ≥98%
Chemical Name or Material 3-Amino-4-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.