missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzoic acid, 99%

Product Code. 11460553
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11460553 5 g 5g
11470553 25 g 25g
11480553 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11460553

Brand: Thermo Scientific Alfa Aesar A12674.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2458-12-0 | C8H9NO2 | 151.165 g/mol

3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-methylbenzoic acid, is a raw material used in synthesis. It is also used as an organic intermediate, Pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2458-12-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 164°C to 168°C
Quantity 5 g
Beilstein 2082449
Solubility Information Very soluble in water.
Formula Weight 151.16
Percent Purity 99%
Chemical Name or Material 3-Amino-4-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.