missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific™

Product Code. 10783003 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
This item is not returnable. View return policy

Product Code. 10783003

Brand: Thermo Scientific Acros 103980050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2458-12-0
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 168.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9NO2
Linear Formula H2NC6H3(CH3)CO2H
Quantity 5 g
Beilstein 14, 487
Molecular Weight (g/mol) 151.16
Formula Weight 151.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.