missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromobenzotrifluoride, 97%

Product Code. 11945231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11945231 5 g 5g
11955231 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945231 Supplier Thermo Scientific Acros Supplier No. 442660050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54962-75-3 | C7H5BrF3N | 240.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54962-75-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
InChI Key HJTLKVYOWNTDPF-UHFFFAOYSA-N
Synonym 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
PubChem CID 2735880
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6970g/mL
Boiling Point 220.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.697
Formula Weight 240.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Amino-5-bromobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-5-bromobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.