missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-mercapto-1,2,4-triazole, 97+%

Product Code. 11462288
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462288 5 g 5g
11472288 25 g 25g
11482288 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462288 Supplier Thermo Scientific Alfa Aesar Supplier No. A15242.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16691-43-3 | C2H4N4S | 116.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
SMILES NC1=NC(=S)NN1

Specifications

Melting Point >300°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 112404
IUPAC Name 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
Formula Weight 116.15
Percent Purity ≥98%
Chemical Name or Material 3-Amino-5-mercapto-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.