missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-o-cresol, 95%, Thermo Scientific™

Product Code. p-55495 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10032640

Brand: Thermo Scientific Acros 209042500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53222-92-7
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007787
InChI Key FLROJJGKUKLCAE-UHFFFAOYSA-N
Synonym 3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
PubChem CID 104448
IUPAC Name 3-amino-2-methylphenol
SMILES CC1=C(C=CC=C1O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 127°C to 133°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 250 mg
Beilstein 13, 579
Formula Weight 123.15
Percent Purity 95%
Chemical Name or Material 3-Amino-o-cresol, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-o-cresol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.