missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic acid, 98%

Product Code. 11375269
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375269 500 g 500g
11365269 100 g 100g
11355269 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375269 Supplier Thermo Scientific Alfa Aesar Supplier No. A19540.36
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-05-8 | C7H7NO2 | 137.138 g/mol

It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as crystalline Intermediate for azo dyes, pharmaceuticals, and X-ray contrast media.

Solubility
Soluble in water at 20°C 5.9g/L. Solubility in hot methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007795
InChI Key XFDUHJPVQKIXHO-UHFFFAOYSA-N
Synonym m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u
PubChem CID 7419
ChEBI CHEBI:42682
IUPAC Name 3-aminobenzoic acid
SMILES C1=CC(=CC(=C1)N)C(=O)O

Specifications

Melting Point 172°C to 179°C
Density 1.51
Flash Point 150°C (302°F)
Quantity 500 g
Beilstein 471603
Merck Index 14,421
Solubility Information Soluble in water at 20°C 5.9g/L. Solubility in hot methanol is almost transparent.
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material 3-Aminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.