missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic acid, 99+%

Product Code. 10744281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10744281 100 g 100g
10636742 25 g 25g
10572551 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10744281 Supplier Thermo Scientific Acros Supplier No. 155851000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-05-8 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007795
InChI Key XFDUHJPVQKIXHO-UHFFFAOYSA-N
Synonym m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u
PubChem CID 7419
ChEBI CHEBI:42682
IUPAC Name 3-aminobenzoic acid
SMILES C1=CC(=CC(=C1)N)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >172°C
Color Beige-Brown to Brown-White
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2NC6H4CO2H
Quantity 100 g
Beilstein 14, 383
Merck Index 15, 416
Solubility Information Solubility in water: 5.9g/L (15°C). Other solubilities: 3g/100g in ethanol (25°C),slightly soluble in ether
Formula Weight 137.14
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 3-Aminobenzoic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Aminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.