missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic acid, 99+%

Product Code. p-55523 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10636742

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-05-8 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007795
InChI Key XFDUHJPVQKIXHO-UHFFFAOYSA-N
Synonym m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u
PubChem CID 7419
ChEBI CHEBI:42682
IUPAC Name 3-aminobenzoic acid
SMILES C1=CC(=CC(=C1)N)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >172°C
Color Beige-Brown to Brown-White
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2NC6H4CO2H
Quantity 25 g
Beilstein 14, 383
Merck Index 15, 416
Solubility Information Solubility in water: 5.9g/L (15°C). Other solubilities: 3g/100g in ethanol (25°C),slightly soluble in ether
Formula Weight 137.14
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 3-Aminobenzoic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.