missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic acid hydrochloride, 98%, Thermo Scientific™

Product Code. 11358145
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358145 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11358145 Supplier Thermo Scientific Alfa Aesar Supplier No. L08407.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15151-51-6
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012973
InChI Key PTTMMENRLMBXGK-UHFFFAOYSA-N
Synonym 3-aminobenzoic acid hydrochloride, 3-aminobenzoic acid, chloride, 3-aminobenzoic acid hcl, acmc-1bs8f, m-aminobenzoic acid hydrochloride
PubChem CID 3084218
IUPAC Name 3-aminobenzoic acid;hydrochloride
SMILES C1=CC(=CC(=C1)N)C(=O)O.Cl

Specifications

Melting Point ∽286°C (decomposition)
Quantity 100 g
Beilstein 3695285
Sensitivity Light sensitive
Formula Weight 173.6
Percent Purity 98%
Chemical Name or Material 3-Aminobenzoic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.