missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocoumarin, 97%, Thermo Scientific™

Product Code. 13525040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13525040 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13525040 Supplier Thermo Scientific Acros Supplier No. 449510010
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1635-31-0
Melting Point 135.0°C to 139.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H7NO2
Quantity 1 g
Synonym 3-aminocoumarin, 3-amino-2h-chromen-2-one, 3-amino-2-benzopyrone, 3-amino-2h-1-benzopyran-2-one, coumarin amine, coumarin monoamine, amino-chromen-2-one, 3-azanylchromen-2-one, acmc-1bous
InChI Key QWZHDKGQKYEBKK-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)N
IUPAC Name 3-aminochromen-2-one
Molecular Weight (g/mol) 161.16
PubChem CID 74217
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 3-Aminocoumarin
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminocoumarin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.