missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenol, 99%

Product Code. 10249263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10249263 5 g Glass bottle 5g
10386590 250 g Glass bottle 250g
10319323 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249263 Supplier Thermo Scientific Acros Supplier No. 146100050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 591-27-5 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-27-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00007786
InChI Key CWLKGDAVCFYWJK-UHFFFAOYSA-N
Synonym m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
PubChem CID 11568
ChEBI CHEBI:28924
IUPAC Name 3-aminophenol
SMILES NC1=CC=CC(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121.0°C to 124.0°C
Color Gray or Pink to White
Boiling Point 164°C (15.0 hPa)
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4OH
Quantity 5 g
Beilstein 13, 401
Merck Index 15, 456
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol, ether and amyl alcohol, slightly soluble in benzene, very slightly soluble in petroleum ether
Formula Weight 109.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Aminophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 3-Aminophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.