missing translation for 'onlineSavingsMsg'
Learn More

3-aminophthalic acid, 95%

Product Code. 10250693 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10250693 1 g Glass bottle 1g
10197972 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10250693 Supplier Thermo Scientific Acros Supplier No. 375750010
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5434-20-8 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5434-20-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00075053
InChI Key WGLQHUKCXBXUDV-UHFFFAOYSA-N
Synonym 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid
PubChem CID 79490
IUPAC Name 3-aminophthalic acid
SMILES C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 170.0°C to 180.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula NH2C6H3(CO2H)2
Quantity 1 g
Formula Weight 181.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophthalic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminophthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.