missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine-4-carboxylic acid, 95%

Product Code. 11432120
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11432120 1 g 1g
15463827 250 mg 250mg
11442120 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11432120

Brand: Thermo Scientific Alfa Aesar H28714.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7579-20-6 | C6H5N2O2 | 137.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7579-20-6
Molecular Formula C6H5N2O2
Molecular Weight (g/mol) 137.12
MDL Number MFCD00137840
InChI Key FYEQKMAVRYRMBL-UHFFFAOYSA-M
Synonym 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure
PubChem CID 459531
IUPAC Name 3-aminopyridine-4-carboxylic acid
SMILES NC1=CN=CC=C1C([O-])=O

Specifications

Melting Point ∽280°C (decomposition)
Quantity 1 g
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 3-Aminopyridine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.