missing translation for 'onlineSavingsMsg'
Learn More

3-Aminorifamycin S (Technical Grade), TRC

Product Code. 30010358
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010358 5 mg 5mg
30011862 2.5 mg 2.5mg
30011595 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010358 Supplier Toronto Research Chemicals Supplier No. TRCA6292735MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51756-80-0
Molecular Formula C37H46N2O12
Molecular Weight (g/mol) 710.77
Synonym 3-Amino-1,4-dideoxy-1,4-dihydro-1,4-dioxorifamycin;
IUPAC Name [(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-26-amino-2,15,17-trihydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23,27,29-tetraoxo-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1(28),2,4,9,19,21,25-heptaen-13-yl] acetate
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4C(=O)C(=C(N)C(=O)c4c3C2=O)NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C37H46N2O12/c1-15-11-10-12-16(2)36(47)39-27-26(38)31(44)23-24(32(27)45)30(43)20(6)34-25(23)35(46)37(8,51-34)49-14-13-22(48-9)17(3)33(50-21(7)40)19(5)29(42)18(4)28(15)41/h10-15,17-19,22,28-29,33,41-43H,38H2,1-9H3,(H,39,47)/b11-10+,14-13+,16-12-/t15-,17+,18+,19+,22-,28-,29+,33+,37-/m0/s1
Formula Weight 710.3051
Chemical Name or Material 3-Aminorifamycin S (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.