missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylacrylic acid, predominantly trans, 97%

Product Code. 11429063
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429063 100 g 100g
11409063 5 g 5g
11419063 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429063 Supplier Thermo Scientific Alfa Aesar Supplier No. A11147.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 583-06-2 | C10H8O3 | 176.171 g/mol

2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-06-2
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00014015
InChI Key PLPDHGOODMBBGN-VOTSOKGWSA-N
Synonym 3-benzoylacrylic acid, benzoylacrylic acid, 4-oxo-4-phenyl-2-butenoic acid, e-4-oxo-4-phenylbut-2-enoic acid, acrylic acid, 3-benzoyl, 2-butenoic acid, 4-oxo-4-phenyl, 4-oxo-4-phenylbut-2-enoic acid, crotonic acid, 4-oxo-4-phenyl, .beta.-benzoylacrylic acid, trans-3-benzoylacrylic acid
PubChem CID 676116
IUPAC Name (E)-4-oxo-4-phenylbut-2-enoic acid
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)O

Specifications

Melting Point 93°C to 97°C
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 1817876
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 3-Benzoylacrylic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.