missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzylamino)propionitrile, 97%, Thermo Scientific™

Product Code. 10235760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235760 25 g Glass bottle 25g
10002280 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235760 Supplier Thermo Scientific Acros Supplier No. 180600250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706-03-6
Molecular Formula C10H13N2
Molecular Weight (g/mol) 161.23
MDL Number MFCD00001955
InChI Key MWTGBAURSCEGSL-UHFFFAOYSA-O
Synonym 3-benzylamino propanenitrile, 3-benzylamino propionitrile, n-benzyl-2-cyanoethylamine, propanenitrile, 3-phenylmethyl amino, a-cyanoethyl benzylamine, beta-cyanoethylbenzylamine, acmc-1bk6i, 3-benzylamino-propionitrile
PubChem CID 69719
IUPAC Name benzyl(2-cyanoethyl)azanium
SMILES N#CCC[NH2+]CC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2NHCH2CH2CN
Refractive Index 1.5298 to 1.5318 (20°C, 589nm)
Quantity 25 g
Formula Weight 160.22
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-(Benzylamino)propionitrile, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.