missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11317006
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317006 50 g 50g
11307006 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317006 Supplier Thermo Scientific Alfa Aesar Supplier No. L06830.18
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6346-05-0
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003386
InChI Key VQVQZFHUXRSRBZ-UHFFFAOYSA-N
Synonym 3-benzyloxy-4-methoxybenzaldehyde, o-benzylisovanillin, benzaldehyde, 4-methoxy-3-phenylmethoxy, benzyl iso-vanillin, 4-methoxy-3-phenylmethoxy benzaldehyde, 3-benzyloxy-4-methoxy-benzaldehyde, benzylisovanillin, pubchem13136, acmc-1b6ff
PubChem CID 80671
IUPAC Name 4-methoxy-3-phenylmethoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1OCC1=CC=CC=C1

Specifications

Melting Point 60°C to 64°C
Quantity 50 g
Beilstein 794627
Sensitivity Air Sensitive
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material 3-Benzyloxy-4-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.