missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzyloxy-4-methoxybenzoic acid, 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Benzyloxy-4-methoxybenzoic acid is used as Pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
3-Benzyloxy-4-methoxybenzoic acid is used as Pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Specifications
| Melting Point | 177°C to 182°C |
| Quantity | 25 g |
| Beilstein | 2660886 |
| Synonym | 3-benzyloxy-4-methoxybenzoic acid, 3-benzyloxy-p-anisic acid, 3-benzyloxy-4-methoxy benzoic acid, 4-methoxy-3-phenylmethoxy benzoic acid, benzoic acid, 4-methoxy-3-phenylmethoxy, acmc-20an9h, 3-benzyloxy-4-methoxy-benzoic acid, benzoic acid, 3-benzyloxy-4-methoxy, benzoic acid,4-methoxy-3-phenylmethoxy |
| Solubility Information | Insoluble in water. |
| IUPAC Name | 3-(benzyloxy)-4-methoxybenzoic acid |
| Formula Weight | 258.28 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 3-Benzyloxy-4-methoxybenzoic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction