missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyaniline, 98%

Product Code. 15422038
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422038 5 g 5g
15412038 1 g 1g
15432038 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422038 Supplier Thermo Scientific Alfa Aesar Supplier No. H55708.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1484-26-0 | C13H13NO | 199.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-26-0
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00007784
InChI Key IGPFOKFDBICQMC-UHFFFAOYSA-N
Synonym 3-benzyloxyaniline, 3-benzyloxy aniline, benzenamine, 3-phenylmethoxy, unii-iab1vda972, 3-benzyloxy-phenylamine, iab1vda972, 3-phenylmethyl oxy aniline, 3-phenylmethoxy phenylamine, 3-benzyloxy benzenamine, 3-benyloxyaniline
PubChem CID 92892
IUPAC Name 3-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)N

Specifications

Melting Point 63°C to 67°C
Color Brown
Quantity 5 g
Formula Weight 199.25
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Benzyloxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.