missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzyl alcohol, 97%, Thermo Scientific™

Product Code. 10577833 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10577833

Brand: Thermo Scientific Acros 181940010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1700-30-7
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00004642
InChI Key AFKLSWIRJUJWKY-UHFFFAOYSA-N
Synonym 3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
PubChem CID 74341
IUPAC Name (3-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47.0°C to 52.0°C
Color Beige to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2OH
Quantity 1 g
Formula Weight 214.26
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Benzyloxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzyloxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.