missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenylhydrazine hydrochloride, 98%

Product Code. 10166913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10166913 5 g Glass bottle 5g
10370774 250 mg Glass bottle 250mg
10390294 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10166913 Supplier Thermo Scientific Acros Supplier No. 394020050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59146-68-8 | C13H15ClN2O | 250.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59146-68-8
Molecular Formula C13H15ClN2O
Molecular Weight (g/mol) 250.73
MDL Number MFCD02258768
InChI Key MMMGCMOISVZVKZ-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylhydrazine hydrochloride, 3-benzyloxy phenyl hydrazine hydrochloride, 3-benzyloxy-phenyl-hydrazine hydrochloride, 3-benzyloxyphenyl hydrazine hydrochloride, 3-benzyloxyphenylhydrazine hcl, benzyloxy-phenyl-hydrazine hydrochloride, 3-phenylmethoxyphenyl diazane hydrochloride, n'-3-benzyloxy-phenyl-hydrazinium, chloride
PubChem CID 12891839
IUPAC Name (3-phenylmethoxyphenyl)hydrazine;hydrochloride
SMILES Cl.NNC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 155.0°C to 157.0°C
Color Violet
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4NHNH2·HCl
Quantity 5 g
Formula Weight 250.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Benzyloxyphenylhydrazine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Benzyloxyphenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.