missing translation for 'onlineSavingsMsg'
Learn More

(3-Benzyloxypropyl)triphenylphosphonium bromide, 97+%

Product Code. 15442276
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442276 25 g 25g
15422276 1 g 1g
15432276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442276 Supplier Thermo Scientific Alfa Aesar Supplier No. H55754.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54314-85-1 | C28H28BrOP | 491.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54314-85-1
Molecular Formula C28H28BrOP
Molecular Weight (g/mol) 491.41
MDL Number MFCD00191781
InChI Key DWYCJWXMZGVGJV-UHFFFAOYSA-M
Synonym 3-benzyloxypropyl triphenylphosphonium bromide, 3-benzyloxy propyl triphenylphosphonium bromide, 3-benzyloxypropyl triphenylphosphoniumbromide, phosphonium, triphenyl 3-phenylmethoxy propyl-, bromide, phosphonium,triphenyl 3-phenylmethoxy propyl-, bromide 1:1, acmc-1asqr, c28h28op.br, 3-benzyloxypropyltriphenylphosphonium bromide, 3-benzyloxypropyl triphenyl phosphonium bromide, 3-benzyloxypropyl triphenyl-phosphonium bromide
PubChem CID 10929077
IUPAC Name triphenyl(3-phenylmethoxypropyl)phosphanium;bromide
SMILES [Br-].C(COCC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C to 154°C
Color White
Quantity 25 g
Formula Weight 491.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (3-Benzyloxypropyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.